2,4,6-Trichlorbenzonitril

Lieferant: FLUOROCHEM

019058-5G
FLUO019058-5GEA 0 EUR
FLUO019058-5G
2,4,6-Trichlorbenzonitril
2,4,6-Trichlorbenzonitril
Formel: C₇H₂Cl₃N
MDL: MFCD00052711
CAS-Nummer: 6575-05-9
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR