2,5-Dibrom-p-xylol

Lieferant: FLUOROCHEM

Synonyme: 2,5-Dibrom-1,4-dimethylbenzol, 1,4-Dibrom-2,5-dimethylbenzol

019177-250G 019177-50G
FLUO019177-250GEA 0 EUR
FLUO019177-250G FLUO019177-50G
2,5-Dibrom-p-xylol
2,5-Dibrom-p-xylol
Formel: C₈H₈Br₂
Molecular Weight: 263.96 g/mol
Siedepunkt: 261
Schmelzpunkt: 75
MDL: MFCD00000091
CAS-Nummer: 1074-24-4
EINECS: 214-038-2
UN: 3077
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR