1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluor-12-ioddodecan

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1H,1H,2H,2H-Perfluorododecyl iodide, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Henicosafluoro-12-iodododecane, 1H,1H,2H,2H-Perfluoro-1-iodododecane

019182-25G
FLUO019182-25GEA 0 EUR
FLUO019182-25G
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluor-12-ioddodecan
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluor-12-ioddodecan
Formel: C₁₂H₄F₂₁I
Molecular Weight: 674,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039412
CAS-Nummer: 2043-54-1
EINECS: 218-054-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR