(±)-2-Piperazincarbonsäure Dihydrochlorid

Lieferant: FLUOROCHEM

Synonyme: Piperazine-2-carboxylic acid dihydrochloride

019229-1G
FLUO019229-1GEA 0 EUR
FLUO019229-1G
(±)-2-Piperazincarbonsäure Dihydrochlorid
(±)-2-Piperazincarbonsäure Dihydrochlorid
Formel: C₅H₁₂Cl₂N₂O₂
Molecular Weight: 203,07 g/mol
MDL: MFCD00044803
CAS-Nummer: 3022-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR