1-(2-Nitrophenyl)piperazin
Lieferant: FLUOROCHEM
019232-1G
FLUO019232-1GEA
31.1
EUR
FLUO019232-1G
1-(2-Nitrophenyl)piperazin
1-(2-Nitrophenyl)piperazin
Formel:
C₁₀H₁₃N₃O₂ Molecular Weight: 207,23 g/mol |
MDL:
MFCD00040728 CAS-Nummer: 59084-06-9 EINECS: 261-593-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...