1-Benzhydrylpiperazin

Lieferant: FLUOROCHEM

Synonyme: 1-(Diphenylmethyl)piperazine

019233-5G
FLUO019233-5GEA 0 EUR
FLUO019233-5G
1-Benzhydrylpiperazin
1-Benzhydrylpiperazin
Formel: C₁₇H₂₀N₂
Molecular Weight: 252.36 g/mol
MDL: MFCD00038379
CAS-Nummer: 841-77-0
EINECS: 212-667-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR