1-(4-tert-Butylbenzyl)piperazin

Lieferant: FLUOROCHEM

019236-1G
FLUO019236-1GEA 0 EUR
FLUO019236-1G
1-(4-tert-Butylbenzyl)piperazin
1-(4-tert-Butylbenzyl)piperazin
Formel: C₁₅H₂₄N₂
Molecular Weight: 232,37 g/mol
MDL: MFCD00082594
CAS-Nummer: 956-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR