1-(4-Fluorbenzyl)piperazin

Lieferant: FLUOROCHEM

019240-1G
FLUO019240-1GEA 0 EUR
FLUO019240-1G
1-(4-Fluorbenzyl)piperazin
1-(4-Fluorbenzyl)piperazin
Formel: C₁₁H₁₅FN₂
Molecular Weight: 194,25 g/mol
MDL: MFCD01075234
CAS-Nummer: 70931-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR