2,6-Dimethylbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dimethylbenzolsulfonsäurechlorid, m-Xylol-2-sulfonsäurechlorid, m-Xylol-2-sulfonylchlorid, 2,6-Dimethylbenzolsulfochlorid, 2,6-Dimethylbenzol-1-sulfonylchlorid

019244-5G
FLUO019244-5GEA 0 EUR
FLUO019244-5G
2,6-Dimethylbenzolsulfonylchlorid
2,6-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Molecular Weight: 204.68 g/mol
MDL: MFCD03789123
CAS-Nummer: 2905-29-5
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR