2,6-Difluorbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 2,6-Difluorobenzohydrazide

019258-1G
FLUO019258-1GEA 179 EUR
FLUO019258-1G
2,6-Difluorbenzhydrazid
2,6-Difluorbenzhydrazid
Formel: C₇H₆F₂N₂O
Molecular Weight: 172,132 g/mol
MDL: MFCD00119406
CAS-Nummer: 172935-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR