(±)-3-Brommandelsäure
Lieferant: FLUOROCHEM
Synonyme:
(±)-2-(3-Bromphenyl)-2-hydroxyessigsäure, (±)-2-(3-Bromphenyl)-2-hydroxyethansäure, (±)-3-Bromphenylglykolsäure
019271-5G
FLUO019271-5GEA
0
EUR
FLUO019271-5G
(±)-3-Brommandelsäure
(±)-3-Brommandelsäure
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol |
MDL:
MFCD03789129 CAS-Nummer: 49839-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...