3-Fluorthioanisol
Lieferant: FLUOROCHEM
Synonyme:
1-Fluor-3-(methylthio)benzol, 3-Fluorphenylmethylsulfid
019273-5G
FLUO019273-5GEA
72.9
EUR
FLUO019273-5G
3-Fluorthioanisol
3-Fluorthioanisol
Formel:
C₇H₇FS Molecular Weight: 142,2 g/mol |
MDL:
MFCD03789131 CAS-Nummer: 658-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...