4',5'-Dimethyl-2'-methoxyacetophenon

Lieferant: FLUOROCHEM

019295-5G
FLUO019295-5GEA 296 EUR
FLUO019295-5G
4',5'-Dimethyl-2'-methoxyacetophenon
4',5'-Dimethyl-2'-methoxyacetophenon
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
MDL: MFCD03789142
CAS-Nummer: 91969-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR