3',5'-Dichlor-2'-methoxyacetophenon

Lieferant: FLUOROCHEM

019296-5G
FLUO019296-5GEA 288 EUR
FLUO019296-5G
3',5'-Dichlor-2'-methoxyacetophenon
3',5'-Dichlor-2'-methoxyacetophenon
Formel: C₉H₈Cl₂O₂
MDL: MFCD03789120
CAS-Nummer: 154260-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR