5'-Ethyl-2'-methoxy-3'-nitroacetophenon

Lieferant: FLUOROCHEM

019299-1G
FLUO019299-1GEA 372 EUR
FLUO019299-1G
5'-Ethyl-2'-methoxy-3'-nitroacetophenon
5'-Ethyl-2'-methoxy-3'-nitroacetophenon
Formel: C₁₁H₁₃NO₄
MDL: MFCD03789146
CAS-Nummer: 685892-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR