4-Fluor-2-iodanilin

Lieferant: FLUOROCHEM

019309-5G 019309-25G
FLUO019309-5GEA 0 EUR
FLUO019309-5G FLUO019309-25G
4-Fluor-2-iodanilin
4-Fluor-2-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237.02 g/mol
MDL: MFCD00042584
CAS-Nummer: 61272-76-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR