(3-Chlorphenyl)methylsulfon

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-3-(methylsulfonyl)benzol

019334-1G
FLUO019334-1GEA 159 EUR
FLUO019334-1G
(3-Chlorphenyl)methylsulfon
(3-Chlorphenyl)methylsulfon
Formel: C₇H₇ClO₂S
Molecular Weight: 190,65 g/mol
CAS-Nummer: 21383-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR