3-(Methylthio)benzotrifluorid
Lieferant: FLUOROCHEM
Synonyme:
Methyl(3-(trifluoromethyl)phenyl)sulphane, 3-Trifluoromethylthioanisole
019342-5G
FLUO019342-5GEA
582
EUR
FLUO019342-5G
3-(Methylthio)benzotrifluorid
3-(Methylthio)benzotrifluorid
Formel:
C₈H₇F₃S Molecular Weight: 192,2014 g/mol |
MDL:
MFCD03789168 CAS-Nummer: 328-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...