3,4-Dichlorthioanisol

Lieferant: FLUOROCHEM

Synonyme: 1,2-Dichlor-4-(methylthio)benzol

019347-10G
FLUO019347-10GEA 0 EUR
FLUO019347-10G
3,4-Dichlorthioanisol
3,4-Dichlorthioanisol
Formel: C₇H₆Cl₂S
Molecular Weight: 193.1 g/mol
CAS-Nummer: 17733-23-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR