3,5-Bis(trifluormethyl)thioanisol

Lieferant: FLUOROCHEM

Synonyme: (3,5-Bis(trifluoromethyl)phenyl)(methyl)sulphane

019353-5G
FLUO019353-5GEA 0 EUR
FLUO019353-5G
3,5-Bis(trifluormethyl)thioanisol
3,5-Bis(trifluormethyl)thioanisol
Formel: C₉H₆F₆S
Molecular Weight: 260,199 g/mol
MDL: MFCD03789174
CAS-Nummer: 685892-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR