2-(Methylthio)benzotrifluorid

Lieferant: FLUOROCHEM

019355-5G
FLUO019355-5GEA 918 EUR
FLUO019355-5G
2-(Methylthio)benzotrifluorid
2-(Methylthio)benzotrifluorid
Formel: C₈H₇F₃S
MDL: MFCD01882714
CAS-Nummer: 322-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR