1,2-Dichlor-4-(methylsulfonyl)benzol
Lieferant: FLUOROCHEM
019370-1G
FLUO019370-1GEA
48.8
EUR
FLUO019370-1G
1,2-Dichlor-4-(methylsulfonyl)benzol
1,2-Dichloro-4-methylsulphonylbenzene
Formel:
C₇H₆Cl₂O₂S |
MDL:
MFCD00025063 CAS-Nummer: 38452-47-0 EINECS: 253-942-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...