4-Amino-1-Boc-piperidin

Lieferant: FLUOROCHEM

019433-1G 019433-5G
FLUO019433-1GEA 31.1 EUR
FLUO019433-1G FLUO019433-5G
4-Amino-1-Boc-piperidin
4-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD01076201
CAS-Nummer: 87120-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR