1-(2-Bromphenyl)piperazin

Lieferant: FLUOROCHEM

019434-1G 019434.10G
FLUO019434-1GEA 0 EUR
FLUO019434-1G FLUO019434.10G
1-(2-Bromphenyl)piperazin
1-(2-Bromphenyl)piperazin
Formel: C₁₀H₁₃BrN₂
Molecular Weight: 241,13 g/mol
MDL: MFCD03844726
CAS-Nummer: 1011-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR