2',4'-Dichlor-5'-fluoracetophenon

Lieferant: FLUOROCHEM

019457-25G
FLUO019457-25GEA 37.4 EUR
FLUO019457-25G
2',4'-Dichlor-5'-fluoracetophenon
2',4'-Dichlor-5'-fluoracetophenon
Formel: C₈H₅Cl₂FO
MDL: MFCD00077499
CAS-Nummer: 704-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR