α-Brom-2,3,5,6-tetrafluortoluol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 2,3,5,6-Tetrafluorbenzylbromid

019466-10G
FLUO019466-10GEA 0 EUR
FLUO019466-10G
α-Brom-2,3,5,6-tetrafluortoluol
α-Brom-2,3,5,6-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
Siedepunkt: 79…80 °C (1013 hPa)
MDL: MFCD00013549
CAS-Nummer: 53001-73-3
EINECS: 258-289-6
UN: 3265
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR