6-Fluornicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-5-pyridinecarboxylic acid

019469-1G
FLUO019469-1GEA 31.1 EUR
FLUO019469-1G
6-Fluornicotinsäure
6-Fluornicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD01859863
CAS-Nummer: 403-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR