3-Chlor-2-fluorphenylisothiocyanat

Lieferant: FLUOROCHEM

019480-1G
FLUO019480-1GEA 249 EUR
FLUO019480-1G
3-Chlor-2-fluorphenylisothiocyanat
3-Chlor-2-fluorphenylisothiocyanat
Formel: C₇H₃ClFNS
CAS-Nummer: 127142-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR