4,5-Dibromveratrol

Lieferant: FLUOROCHEM

Synonyme: 1,2-Dibromo-4,5-dimethoxybenzene, 4,5-Dibromoveratrole

019483-25G
FLUO019483-25GEA 173 EUR
FLUO019483-25G
4,5-Dibromveratrol
4,5-Dibromveratrol
Formel: C₈H₈Br₂O₂
MDL: MFCD00014894
CAS-Nummer: 37895-73-1
EINECS: 253-705-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR