Methyl-2-chlor-6-methylisonicotinat

Lieferant: FLUOROCHEM

Synonyme: Methyl 2-chloro-6-picoline-4-carboxylate, Methyl 2-chloro-6-methylpyridine-4-carboxylate

019484-2.5G
FLUO019484-2.5GEA 0 EUR
FLUO019484-2.5G
Methyl-2-chlor-6-methylisonicotinat
Methyl-2-chlor-6-methylisonicotinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
MDL: MFCD04038391
CAS-Nummer: 3998-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR