2,4-Difluor-5-nitroanilin

Lieferant: FLUOROCHEM

019497-1G
FLUO019497-1GEA 63.9 EUR
FLUO019497-1G
2,4-Difluor-5-nitroanilin
2,4-Difluor-5-nitroanilin
Formel: C₆H₄F₂N₂O₂
Molecular Weight: 174,11 g/mol
MDL: MFCD04038395
CAS-Nummer: 123344-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR