1,3-Difluor-5-nitrobenzol

Lieferant: FLUOROCHEM
Achtung

019498-5G
FLUO019498-5GEA 51.8 EUR
FLUO019498-5G
1,3-Difluor-5-nitrobenzol
1,3-Difluor-5-nitrobenzol
Formel: C₆H₃F₂NO₂
Molecular Weight: 159,09 g/mol
MDL: MFCD00012142
CAS-Nummer: 2265-94-3
EINECS: 218-867-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR