Diethyl(difluormethyl)phosphonat

Lieferant: FLUOROCHEM

Synonyme: Difluoromethylphosphonicaciddiethylester

020007-1G
FLUO020007-1GEA 302 EUR
FLUO020007-1G
Diethyl(difluormethyl)phosphonat
Diethyl(difluormethyl)phosphonat
Formel: C₅H₁₁F₂O₃P
Molecular Weight: 188,1096 g/mol
MDL: MFCD00192023
CAS-Nummer: 1478-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR