α-(Trifluormethyl)-4,4'-bis(methoxy)benzhydrol
Lieferant: FLUOROCHEM
020036-1G
FLUO020036-1GEA
200
EUR
FLUO020036-1G
α-(Trifluormethyl)-4,4'-bis(methoxy)benzhydrol
α-(Trifluormethyl)-4,4'-bis(methoxy)benzhydrol
Formel:
C₁₆H₁₅F₃O₃ |
MDL:
MFCD04116451 CAS-Nummer: 379-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...