1-Fluor-3-iod-5-nitrobenzol

Lieferant: FLUOROCHEM

020051-1G 020051.5G
FLUO020051-1GEA 104 EUR
FLUO020051-1G FLUO020051.5G
1-Fluor-3-iod-5-nitrobenzol
1-Fluoro-3-iodo-5-nitrobenzene
Formel: C₆H₃FINO₂
MDL: MFCD00007219
CAS-Nummer: 3819-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR