3-Fluor-5-iodanilin

Lieferant: FLUOROCHEM

020052-1G
FLUO020052-1GEA 84.9 EUR
FLUO020052-1G
3-Fluor-5-iodanilin
3-Fluor-5-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
MDL: MFCD04116466
CAS-Nummer: 660-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR