3-Fluor-5-iodbenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 5-Fluoro-3-iodobenzoic acid

020053-1G
FLUO020053-1GEA 192 EUR
FLUO020053-1G
3-Fluor-5-iodbenzoesäure
3-Fluor-5-iodbenzoesäure
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
MDL: MFCD04116467
CAS-Nummer: 723294-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR