3-Fluor-5-iodbenzonitril

Lieferant: FLUOROCHEM

020054-1G
FLUO020054-1GEA 0 EUR
FLUO020054-1G
3-Fluor-5-iodbenzonitril
3-Fluor-5-iodbenzonitril
Formel: C₇H₃FIN
Molecular Weight: 247,0100 g/mol
MDL: MFCD04116468
CAS-Nummer: 723294-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR