Ethyl-6-brompicolinat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 6-bromopyridine-2-carboxylate
020124-5G
FLUO020124-5GEA
45.9
EUR
FLUO020124-5G
Ethyl-6-brompicolinat
Ethyl-6-brompicolinat
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,0600 g/mol |
MDL:
MFCD04116913 CAS-Nummer: 21190-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...