6-Aminonicotinonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-5-pyridincarbonsäurenitril, 2-Amino-5-cyanpyridin, 6-Aminonicotinsäurenitril, 2-Amino-5-pyridincarbonitril

020194-1G
FLUO020194-1GEA 0 EUR
FLUO020194-1G
6-Aminonicotinonitril
6-Aminonicotinonitril
Formel: C₆H₅N₃
Molecular Weight: 119,13 g/mol
Siedepunkt: 240…250 °C (15 mmHg)
Schmelzpunkt: 164 °C
MDL: MFCD00128902
CAS-Nummer: 4214-73-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR