4'-Formylbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Formyl-[1,1'-biphenyl]-3-carboxylic acid

020320-1G
FLUO020320-1GEA 206 EUR
FLUO020320-1G
4'-Formylbiphenyl-3-carbonsäure
4'-Formylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₀O₃
Molecular Weight: 226,2274 g/mol
MDL: MFCD03426484
CAS-Nummer: 222180-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR