3'-Methylbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3'-Methyl[1,1'-biphenyl]-3-carboxylic acid

020321-1G
FLUO020321-1GEA 82.1 EUR
FLUO020321-1G
3'-Methylbiphenyl-3-carbonsäure
3'-Methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
MDL: MFCD00558892
CAS-Nummer: 158619-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR