2,6-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
Lieferant: FLUOROCHEM
CFF1056.1
CFF1056.5
CFF1056.25
020624-1G
CHMPCFF1056.1EA
522
EUR
CHMPCFF1056.1
CHMPCFF1056.5
CHMPCFF1056.25
FLUO020624-1G
2,6-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
2,6-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel:
C₁₀H₁₀N₂O |
MDL:
MFCD04117274 CAS-Nummer: 728864-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...