Ethylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: FLUOROCHEM

020734-1G
FLUO020734-1GEA 34.1 EUR
FLUO020734-1G
Ethylimidazo[1,2-a]pyridin-2-carboxylat
Ethylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₁₀N₂O₂
MDL: MFCD02325394
CAS-Nummer: 38922-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR