1-(3,4-Dimethylphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Gefahr

020737-1G
FLUO020737-1GEA 713 EUR
FLUO020737-1G
1-(3,4-Dimethylphenyl)-2-thioharnstoff
1-(3,4-Dimethylphenyl)-2-thioharnstoff
Formel: (C₆H₃(CH₃)₂)NC(S)NH₂
Molecular Weight: 180,27 g/mol
MDL: MFCD00060452
CAS-Nummer: 16738-18-4
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR