1-(2,5-Dimethylphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
020738-1G
FLUO020738-1GEA
282
EUR
FLUO020738-1G
1-(2,5-Dimethylphenyl)-2-thioharnstoff
1-(2,5-Dimethylphenyl)-2-thioharnstoff
Formel:
(C₆H₃(CH₃)₂)NC(S)NH₂ Molecular Weight: 180,27 g/mol |
MDL:
MFCD00041372 CAS-Nummer: 16738-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...