N-(6-Methyl-2-pyridyl)thioharnstoff
Lieferant: FLUOROCHEM
020742-1G
FLUO020742-1GEA
170
EUR
FLUO020742-1G
N-(6-Methyl-2-pyridyl)thioharnstoff
N-(6-Methyl-2-pyridyl)thioharnstoff
Formel:
C₇H₉N₃S Molecular Weight: 167,23 g/mol |
MDL:
MFCD00462739 CAS-Nummer: 49600-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...