N-(5-Methyl-2-pyridyl)thioharnstoff
Lieferant: FLUOROCHEM
020743-1G
FLUO020743-1GEA
76.2
EUR
FLUO020743-1G
N-(5-Methyl-2-pyridyl)thioharnstoff
N-(5-Methyl-2-pyridyl)thioharnstoff
Formel:
C₇H₉N₃S Molecular Weight: 167,23 g/mol |
MDL:
MFCD04117347 CAS-Nummer: 131185-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...