N-(4-Methyl-2-pyridyl)thioharnstoff
Lieferant: FLUOROCHEM
020744-1G
FLUO020744-1GEA
971
EUR
FLUO020744-1G
N-(4-Methyl-2-pyridyl)thioharnstoff
N-(4-Methyl-2-pyridyl)thioharnstoff
Formel:
C₇H₉N₃S |
MDL:
MFCD04117348 CAS-Nummer: 21242-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...