N-(3-Methyl-2-pyridyl)thioharnstoff

Lieferant: FLUOROCHEM

020745-1G
FLUO020745-1GEA 381 EUR
FLUO020745-1G
N-(3-Methyl-2-pyridyl)thioharnstoff
N-(3-Methyl-2-pyridyl)thioharnstoff
Formel: C₇H₉N₃S
Molecular Weight: 167,23 g/mol
MDL: MFCD03617970
CAS-Nummer: 41440-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR